Gene Transcripts
- Gene Symbol
- Nxt2
- Entrez Gene ID
- 237082
- Gene Name
- nuclear transport factor 2-like export factor 2
- Gene Aliases
- 6330587F24Rik, AI787442, P15-2
- Chromosome Location
- Chr. X: 142226770 - 142239700 on Build GRCm38
- UniGene ID
- Mm.41899
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161430.2 |
NP_001154902.1
|
3, 4 |
260 |
|
NM_001290532.1 |
NP_001277461.1
|
3, 4 |
345 |
|
NM_001290533.1 |
NP_001277462.1
|
2, 3 |
351 |
|
NM_172782.4 |
NP_766370.2
|
2, 3 |
297 |
|
GenBank mRNA |
AK032102.1 |
- |
2, 3 |
268 |
|
AK144349.1 |
- |
2, 3 |
281 |
|
BB851047.1 |
- |
2, 3 |
299 |
|
BB857403.1 |
- |
3, 4 |
261 |
|
BC064727.1 |
- |
2, 3 |
272 |
|
BC068166.1 |
- |
2, 3 |
250 |
30361776 |
BC094570.1 |
- |
2, 3 |
214 |
30361291 |
BC118503.1 |
- |
2, 3 |
225 |
40106658 |
CA482101.1 |
- |
3, 4 |
296 |
|
CJ148018.1 |
- |
3, 4 |
346 |
|
View Full Product Details