Gene Transcripts
- Gene Symbol
- Nxt2
- Entrez Gene ID
- 237082
- Gene Name
- nuclear transport factor 2-like export factor 2
- Gene Aliases
- 6330587F24Rik, AI787442, P15-2
- Chromosome Location
- Chr. X: 142226770 - 142239700 on Build GRCm38
- UniGene ID
- Mm.41899
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK032102.1 |
- |
4 |
543 |
|
View Full Product Details