Gene Transcripts

Gene Symbol
Neu2
Entrez Gene ID
23956
Gene Name
neuraminidase 2
Gene Aliases
MBS, MSS, MTS
Chromosome Location
Chr. 1: 87574027 - 87597845 on Build GRCm38
UniGene ID
Mm.143717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160163.1 NP_001153635.1 4 1037
NM_001160164.1 NP_001153636.1 3 935
NM_001160165.1 NP_001153637.1 3 1063
NM_015750.3 NP_056565.1 3 903
GenBank mRNA AB028023.1 - 3 905
AB048604.1 - 3 1160
AF139059.1 - 3 1014
AK009828.1 - 2 1095
AK145284.1 - 3 936
BC026408.1 - 2 691
BC105657.1 - 3 905 40046586
BC105658.1 - 3 905 40046587
BC105659.1 - 3 905 40046588
BC105661.1 - 2 1165 40046584

View Full Product Details