Gene Transcripts

Gene Symbol
Ptchd2
Entrez Gene ID
242748
Gene Name
patched domain containing 2
Gene Aliases
G630052C06Rik, RNDEu-2
Chromosome Location
Chr. 4: 148236433 - 148288161 on Build GRCm38
UniGene ID
Mm.33873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083342.1 NP_001076811.1 18 3524
GenBank mRNA AB235901.1 - 17 3278
AK078196.1 - 8 1023
AK090326.1 - 18 3547
AK173152.1 - 17 2967
BC070441.1 - 18 3398 30535318
BC072569.1 - 18 3423 5719275

View Full Product Details