Gene Transcripts

Gene Symbol
Tshz3
Entrez Gene ID
243931
Gene Name
teashirt zinc finger family member 3
Gene Aliases
A630038G13Rik, Tsh3, Zfp537, mKIAA1474, teashirt3
Chromosome Location
Chr. 7: 36697742 - 36773553 on Build GRCm38
UniGene ID
Mm.44141
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172298.2 NP_758502.1 2 2885
GenBank mRNA AK037176.1 - 1 208
AK041793.1 - 1 1771
AK086984.1 - 1 1268
AK220396.1 - 2 2756
AY063491.1 - 2 2885
BC150674.1 - 2 2828 9087585

View Full Product Details