Gene Transcripts

Gene Symbol
Il17f
Entrez Gene ID
257630
Gene Name
interleukin 17F
Gene Aliases
C87042, IL-17F
Chromosome Location
Chr. 1: 20777146 - 20785274 on Build GRCm38
UniGene ID
Mm.222807
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145856.2 NP_665855.2 2 271
GenBank mRNA AB116259.1 - 2 200
AF458064.1 - 1 176
AY380822.1 - 2 271
BC145977.1 - 1 216

View Full Product Details