Gene Transcripts
- Gene Symbol
- Larp7
- Entrez Gene ID
- 28036
- Gene Name
- La ribonucleoprotein domain family, member 7
- Gene Aliases
- C330027G06Rik, D3Wsu161e
- Chromosome Location
- Chr. 3: 127536714 - 127553349 on Build GRCm38
- UniGene ID
- Mm.291032
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138593.2 |
NP_613059.2
|
8, 9 |
1305 |
|
GenBank mRNA |
BM946683.1 |
- |
2, 3 |
173 |
|
View Full Product Details