Gene Transcripts

Gene Symbol
Vwc2l
Entrez Gene ID
320460
Gene Name
von Willebrand factor C domain-containing protein 2-like
Gene Aliases
A830006F12Rik, Brl
Chromosome Location
Chr. 1: 70725221 - 70886514 on Build GRCm38
UniGene ID
Mm.360649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177164.3 NP_796138.2 2 889
GenBank mRNA AB374230.1 - 1 85
AK080585.1 - 3 479
AK083856.1 - 2 853
BC064448.1 - 2 702
BC094541.1 - 2 889 6410144
EF552208.1 - 1 153
EF552209.1 - 1 88
EF552210.1 - 1 88

View Full Product Details