Gene Transcripts
- Gene Symbol
- Arl9
- Entrez Gene ID
- 384185
- Gene Name
- ADP-ribosylation factor-like 9
- Gene Aliases
- 1700110A09Rik
- Chromosome Location
- Chr. 5: 77002805 - 77010959 on Build GRCm38
- UniGene ID
- Mm.451858
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_206935.1 |
NP_996818.1
|
2 |
133 |
|
- Gene Symbol
- 1700112J05Rik
- Entrez Gene ID
- 68246
- Gene Name
- RIKEN cDNA 1700112J05 gene
- Gene Aliases
- 1700110A09Rik
- Chromosome Location
- Chr. 5: 77006442 - 77018863 on Build GRCm38
- UniGene ID
- Mm.319633
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018948.1 |
- |
4 |
680 |
|
AV206224.1 |
- |
1 |
239 |
|
View Full Product Details