Gene Transcripts
- Gene Symbol
- Postn
- Entrez Gene ID
- 50706
- Gene Name
- periostin, osteoblast specific factor
- Gene Aliases
- A630052E07Rik, AI747096, OSF-2, Osf2, PLF, PN, peri
- Chromosome Location
- Chr. 3: 54361096 - 54391041 on Build GRCm38
- UniGene ID
- Mm.236067
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198765.1 |
NP_001185694.1
|
3, 4 |
333 |
|
NM_001198766.1 |
NP_001185695.1
|
3, 4 |
333 |
|
NM_015784.3 |
NP_056599.1
|
3, 4 |
333 |
|
GenBank mRNA |
AK030756.1 |
- |
3, 4 |
332 |
|
AK154049.1 |
- |
3, 4 |
331 |
|
AY651928.1 |
- |
3, 4 |
303 |
|
BC007141.1 |
- |
3, 4 |
326 |
|
BC031449.1 |
- |
3, 4 |
302 |
4457222 |
D13664.1 |
- |
3, 4 |
315 |
|
E05655.1 |
- |
3, 4 |
329 |
|
KM117171.1 |
- |
3, 4 |
297 |
|
KM117172.1 |
- |
3, 4 |
297 |
|
View Full Product Details