Gene Transcripts

Gene Symbol
Rgs1
Entrez Gene ID
50778
Gene Name
regulator of G-protein signaling 1
Gene Aliases
BL34
Chromosome Location
Chr. 1: 144244669 - 144249104 on Build GRCm38
UniGene ID
Mm.103701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015811.2 NP_056626.2 4 418
GenBank mRNA AF215667.1 - 4 349
AK089293.1 - 3 885
AK138688.1 - 4 374
AK154841.1 - 2774
AK170366.1 - 4 370
AK170957.1 - 4 374
AK171306.1 - 715
BC028634.1 - 4 349 1328269
BC052684.1 - 2 1204

View Full Product Details