Gene Transcripts

Gene Symbol
Lgi1
Entrez Gene ID
56839
Gene Name
leucine-rich repeat LGI family, member 1
Gene Aliases
BB130740
Chromosome Location
Chr. 19: 38264782 - 38308939 on Build GRCm38
UniGene ID
Mm.298251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020278.2 NP_064674.1 1 242
GenBank mRNA AK031996.1 - 1 144
AK035873.1 - 1 212
AK045443.1 - 1 347
BC066090.1 - 1 242 6839091

View Full Product Details