Gene Transcripts
- Gene Symbol
- Dolpp1
- Entrez Gene ID
- 57170
- Gene Name
- dolichyl pyrophosphate phosphatase 1
- Gene Aliases
- 0610011H20Rik, AB030189, LSFR2
- Chromosome Location
- Chr. 2: 30392124 - 30400529 on Build GRCm38
- UniGene ID
- Mm.285167
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290508.1 |
NP_001277437.1
|
8, 9 |
1071 |
|
NM_001290509.1 |
NP_001277438.1
|
6, 7 |
735 |
|
NM_020329.4 |
NP_065062.1
|
7, 8 |
864 |
|
GenBank mRNA |
AB030189.1 |
- |
7, 8 |
704 |
|
AK002544.1 |
- |
8, 9 |
910 |
|
AK010797.1 |
- |
3, 4 |
301 |
|
AK044141.1 |
- |
7, 8 |
703 |
|
AK053923.1 |
- |
7, 8 |
693 |
|
AK088065.1 |
- |
7, 8 |
709 |
|
BC026544.1 |
- |
7, 8 |
688 |
4975127 |
View Full Product Details