Gene Transcripts

Gene Symbol
Arhgef26
Entrez Gene ID
622434
Gene Name
Rho guanine nucleotide exchange factor (GEF) 26
Gene Aliases
4631416L12Rik, 8430436L14Rik, AI303572, AW491953, SGEF
Chromosome Location
Chr. 3: 62338777 - 62462221 on Build GRCm38
UniGene ID
Mm.153974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081295.1 NP_001074764.1 5 2097
GenBank mRNA AK144976.1 - 5 1603

View Full Product Details