Gene Transcripts

Gene Symbol
Syndig1l
Entrez Gene ID
627191
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
DSPC1, Gm261, Tmem90a
Chromosome Location
Chr. 12: 84677277 - 84698907 on Build GRCm38
UniGene ID
Mm.152484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033334.2 NP_001028506.1 2 358
GenBank mRNA AK140195.1 - 2 358
AY993933.1 - 3 407
AY993934.1 - 2 382
BC132221.1 - 1 75 40130498
BC138501.1 - 1 75 8861521

View Full Product Details