Gene Transcripts

Gene Symbol
Wfdc11
Entrez Gene ID
629761
Gene Name
WAP four-disulfide core domain 11
Gene Aliases
Chromosome Location
Chr. 2: 164662894 - 164674253 on Build GRCm38
UniGene ID
Mm.389336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161806.1 NP_001155278.1 3, 4 304
GenBank mRNA AK079019.1 - 3, 4 305

View Full Product Details