Gene Transcripts

Gene Symbol
Isoc1
Entrez Gene ID
66307
Gene Name
isochorismatase domain containing 1
Gene Aliases
2610034N03Rik
Chromosome Location
Chr. 18: 58659482 - 58679570 on Build GRCm38
UniGene ID
Mm.182574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025478.3 NP_079754.2 4 743
GenBank mRNA AK010892.1 - 1 17
AK011674.1 - 4 610
AK134794.1 - 4 743
AK152763.1 - 4 740
AK153257.1 - 4 740
AK165542.1 - 4 764
AY035213.1 - 4 743
BC016576.1 - 4 750 2609919

View Full Product Details