Gene Transcripts
- Gene Symbol
- Riiad1
- Entrez Gene ID
- 66353
- Gene Name
- regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
- Gene Aliases
- 2310007A19Rik, Ncrna00166
- Chromosome Location
- Chr. 3: 94464984 - 94475078 on Build GRCm38
- UniGene ID
- Mm.46405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025506.2 |
NP_079782.1
|
2, 3 |
167 |
|
GenBank mRNA |
AK009192.1 |
- |
2, 3 |
169 |
|
View Full Product Details