Gene Transcripts

Gene Symbol
Riiad1
Entrez Gene ID
66353
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Gene Aliases
2310007A19Rik, Ncrna00166
Chromosome Location
Chr. 3: 94464984 - 94475078 on Build GRCm38
UniGene ID
Mm.46405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025506.2 NP_079782.1 5, 6 381
GenBank mRNA AK009192.1 - 5, 6 383
AK019569.1 - 3, 4 191
BC107021.1 - 4, 5 768 40049310

View Full Product Details