Gene Transcripts

Gene Symbol
Sar1b
Entrez Gene ID
66397
Gene Name
SAR1 gene homolog B (S. cerevisiae)
Gene Aliases
2310075M17Rik, 2900019I22Rik, CMRD, Sara1b, Sara2, Sarb
Chromosome Location
Chr. 11: 51763663 - 51791953 on Build GRCm38
UniGene ID
Mm.196592
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025535.2 NP_079811.1 2 160
GenBank mRNA AK002327.1 - 2 137
AK010187.1 - 2 143
AK013559.1 - 2 122
AK150906.1 - 2 116
AK155429.1 - 2 130
BC082550.1 - 2 129 30357800
BC085178.1 - 2 113 30701201
BY068863.1 - 2 160

View Full Product Details