Gene Transcripts

Gene Symbol
Nova1
Entrez Gene ID
664883
Gene Name
neuro-oncological ventral antigen 1
Gene Aliases
9430099M15Rik, G630039L02, Nova-1
Chromosome Location
Chr. 12: 46694517 - 46819194 on Build GRCm38
UniGene ID
Mm.247195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021361.1 NP_067336.1 2 229
GenBank mRNA AF232828.1 - 2 163
AK090295.1 - 2 518
BM942596.1 - 2 229

View Full Product Details