Gene Transcripts
- Gene Symbol
- Ubxn6
- Entrez Gene ID
- 66530
- Gene Name
- UBX domain protein 6
- Gene Aliases
- 1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2
- Chromosome Location
- Chr. 17: 56068249 - 56074989 on Build GRCm38
- UniGene ID
- Mm.28000
- Species
- Mus musculus
- Gene Symbol
- Chaf1a
- Entrez Gene ID
- 27221
- Gene Name
- chromatin assembly factor 1, subunit A (p150)
- Gene Aliases
- AL023013, AL024058, CAF-1, Cac1p
- Chromosome Location
- Chr. 17: 56040416 - 56068026 on Build GRCm38
- UniGene ID
- Mm.391010
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132551.1 |
- |
|
6358 |
|
View Full Product Details