Gene Transcripts

Gene Symbol
Snx31
Entrez Gene ID
66696
Gene Name
sorting nexin 31
Gene Aliases
4631426E05Rik
Chromosome Location
Chr. 15: 36504062 - 36555614 on Build GRCm38
UniGene ID
Mm.253627
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025712.4 NP_079988.3 6 565
GenBank mRNA AK014536.1 - 6 567
AK034196.1 - 6 567
AK035226.1 - 6 566
BC061007.1 - 6 561 30314775

View Full Product Details