Gene Transcripts

Gene Symbol
Cep19
Entrez Gene ID
66994
Gene Name
centrosomal protein 19
Gene Aliases
1500031L02Rik, AI428934, AL022620
Chromosome Location
Chr. 16: 32099802 - 32108054 on Build GRCm38
UniGene ID
Mm.22990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025892.2 NP_080168.1 4 599
GenBank mRNA AK005324.1 - 4 599
AK007713.1 - 4 583
BC023715.1 - 3 279 5346460

View Full Product Details