Gene Transcripts

Gene Symbol
Mxra7
Entrez Gene ID
67622
Gene Name
matrix-remodelling associated 7
Gene Aliases
1810057P16Rik, E130302J09Rik
Chromosome Location
Chr. 11: 116803400 - 116828066 on Build GRCm38
UniGene ID
Mm.442120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026280.3 NP_080556.1 4 529
GenBank mRNA AK012601.1 - 4 501
AK075801.1 - 4 530
BC147320.1 - 4 610 9056373
BC147321.1 - 4 610 9056376

View Full Product Details