Gene Transcripts

Gene Symbol
Loh12cr1
Entrez Gene ID
67774
Gene Name
loss of heterozygosity, 12, chromosomal region 1 homolog (human)
Gene Aliases
5830457J20Rik, AW124643
Chromosome Location
Chr. 6: 134639512 - 134711184 on Build GRCm38
UniGene ID
Mm.41892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026371.3 NP_080647.2 1, 2 318
GenBank mRNA AK007423.1 - 1, 2 175
AK018019.1 - 1, 2 234
AK089648.1 - 1, 2 182
AK151342.1 - 1, 2 184
AK169615.1 - 1, 2 190
BC023059.1 - 1, 2 277 5355560
BY077254.1 - 1, 2 321

View Full Product Details