Gene Transcripts

Gene Symbol
Klk5
Entrez Gene ID
68668
Gene Name
kallikrein related-peptidase 5
Gene Aliases
1110030O19Rik
Chromosome Location
Chr. 7: 43834157 - 43851181 on Build GRCm38
UniGene ID
Mm.458122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026806.2 NP_081082.1 4 725
GenBank mRNA AK003996.1 - 4 726
BC139475.1 - 4 726 8862497
BC139478.1 - 4 726 8862500

View Full Product Details