Gene Transcripts

Gene Symbol
Mypn
Entrez Gene ID
68802
Gene Name
myopalladin
Gene Aliases
1110056A04Rik, 9830112J03, AI853556, mKIAA4170
Chromosome Location
Chr. 10: 63115795 - 63203952 on Build GRCm38
UniGene ID
Mm.259767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182992.2 NP_892037.2 11 2216
GenBank mRNA AK036458.1 - 11 2177
AK220521.1 - 11 2216

View Full Product Details