Gene Transcripts

Gene Symbol
Ten1
Entrez Gene ID
69535
Gene Name
TEN1 telomerase capping complex subunit
Gene Aliases
2310004N24Rik
Chromosome Location
Chr. 11: 116198855 - 116215318 on Build GRCm38
UniGene ID
Mm.390347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027107.1 NP_081383.1 3 378
GenBank mRNA AK009156.1 - 3 345
AK146257.1 - 3 368
AK153854.1 - 2 967
BC027639.1 - 3 323 1069092

View Full Product Details