Gene Transcripts
- Gene Symbol
- Ndufa11
- Entrez Gene ID
- 69875
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11
- Gene Aliases
- 2010012C24Rik, AV006275, B14.7
- Chromosome Location
- Chr. 17: 56717762 - 56724248 on Build GRCm38
- UniGene ID
- Mm.279823
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027244.1 |
NP_081520.1
|
2, 3 |
265 |
|
GenBank mRNA |
AK008201.1 |
- |
2, 3 |
260 |
|
View Full Product Details