Gene Transcripts

Gene Symbol
Qpct
Entrez Gene ID
70536
Gene Name
glutaminyl-peptide cyclotransferase (glutaminyl cyclase)
Gene Aliases
5730422A13Rik
Chromosome Location
Chr. 17: 79051906 - 79090268 on Build GRCm38
UniGene ID
Mm.293870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027455.2 NP_081731.1 1 433
GenBank mRNA AK017598.1 - 1 96
AK037197.1 - 1 86
AK045974.1 - 1 434
BC020023.1 - 1 151 3491756

View Full Product Details