Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001035854.2 | NP_001030931.1 | 8 | 1194 | |
NM_027915.3 | NP_082191.1 | 8 | 1194 | ||
GenBank mRNA | AK004975.1 | - | 8 | 1118 | |
AK030505.1 | - | 8 | 1188 | ||
AK032617.1 | - | 8 | 1157 | ||
AK138434.1 | - | 8 | 1195 | ||
BC013699.1 | - | 4 | 580 | ||
BC046772.1 | - | 8 | 1166 | 5708062 |