Gene Transcripts

Gene Symbol
Nkiras2
Entrez Gene ID
71966
Gene Name
NFKB inhibitor interacting Ras-like protein 2
Gene Aliases
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2
Chromosome Location
Chr. 11: 100622955 - 100627602 on Build GRCm38
UniGene ID
Mm.274734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028024.2 NP_082300.1 3 359
GenBank mRNA AK010371.1 - 3 359
AK015918.1 - 364
AK052645.1 - 3 359
AK083475.1 - 3 402
AK149696.1 - 3 503
AK169878.1 - 3 384
BC013469.1 - 2 312 4219892
BC024398.1 - 2 302 5003001

View Full Product Details