Gene Transcripts

Gene Symbol
Cxx1c
Entrez Gene ID
72865
Gene Name
CAAX box 1C
Gene Aliases
2900027G03Rik, Mar8.1, Mart8a
Chromosome Location
Chr. X: 53607922 - 53609135 on Build GRCm38
UniGene ID
Mm.158897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028375.3 NP_082651.1 1 397
GenBank mRNA AK013603.1 - 1 288
AK214233.1 - 1 54
BC031701.1 - 1 285 3594379
CK627323.1 - 1 314

View Full Product Details