Gene Transcripts

Gene Symbol
Ly6k
Entrez Gene ID
76486
Gene Name
lymphocyte antigen 6 complex, locus K
Gene Aliases
2410015A16Rik, 3110035B01Rik, mLy-6K
Chromosome Location
Chr. 15: 74796874 - 74799968 on Build GRCm38
UniGene ID
Mm.273319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029627.2 NP_083903.1 4 560
GenBank mRNA AK010485.1 - 4 564
BC049723.1 - 3 399 6743692

View Full Product Details