Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
79043
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
Chromosome Location
Chr. 17: 24886349 - 24892147 on Build GRCm38
UniGene ID
Mm.274794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027141.2 NP_081417.2 1 84
GenBank mRNA AF403038.1 - 1 84
AK009325.1 - 1 84

View Full Product Details