Product Details
- SNP ID
-
rs13482435
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:12367904 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGGAAGGAGGGCCTGGGCTGGTC[A/G]TTCCCTTTCTTGATAATCATGAGGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pdzd2
PubMed Links
Gene Details
- Gene
- Pdzd2
- Gene Name
- PDZ domain containing 2
View Full Product Details