Product Details

SNP ID
rs13481126
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:83283510 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATTGGCAAATGCAGAAACATAATG[A/T]GCCAGTGTCTCTTTAATCCGAGGGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AA465934 PubMed Links

Gene Details

Gene
AA465934
Gene Name
expressed sequence AA465934
There are no transcripts associated with this gene.

Gene
AI450353
Gene Name
expressed sequence AI450353
There are no transcripts associated with this gene.

Gene
Slfn14
Gene Name
schlafen 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166028.1 782 Silent Mutation NP_001159500.1

View Full Product Details