Product Details
- SNP ID
-
rs13481388
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:34761873 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATGGTGCCCAGCTTTCTCTACTC[C/T]ACCTGCTAAACAATGCAGCAAGAAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Hdac9
PubMed Links
Gene Details
- Gene
- Hdac9
- Gene Name
- histone deacetylase 9
View Full Product Details