Product Details
- SNP ID
-
rs13475761
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:22317036 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTTCTGGCTGTTGCTGCTTTCAA[A/G]ACCAGGTACATGAACAAGGATGGGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rims1
PubMed Links
Gene Details
- Gene
- Rims1
- Gene Name
- regulating synaptic membrane exocytosis 1
There are no transcripts associated with this gene.
View Full Product Details