Product Details

SNP ID
rs27054797
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:105809350 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTTTTCTTTTGCTGTATTTAT[A/G]CCAGTCTTCAGTGATAAGTTGGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Tanc2 PubMed Links

Gene Details

Gene
Tanc2
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181071.3 Intron NP_851416.2

View Full Product Details