Product Details

SNP ID
rs13481227
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:110449131 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCTATCCCTCTTCTCTGTAGGATC[C/T]TTGCAAAGGTGGATTCTTCCGGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Map2k6 PubMed Links

Gene Details

Gene
Map2k6
Gene Name
mitogen-activated protein kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011943.2 Intron NP_036073.1

View Full Product Details