Product Details

SNP ID
rs13482487
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:25951940 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGACATTAGAGAGAAAGGGGATTGT[A/G]TTCGTGGCACTCTTTTTTTCTTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Fam134b PubMed Links

Gene Details

Gene
Fam134b
Gene Name
family with sequence similarity 134, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034851.2 Intron NP_001030023.1
NM_001277315.1 Intron NP_001264244.1
NM_001277316.1 Intron NP_001264245.1
NM_001277317.1 Intron NP_001264246.1
NM_001277318.1 Intron NP_001264247.1
NM_025459.3 Intron NP_079735.2
Gene
Mir7212
Gene Name
microRNA 7212
There are no transcripts associated with this gene.

View Full Product Details