Product Details

SNP ID
rs13475955
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:83338931 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCATAAGAGCTAGGTGATTTCAAT[C/T]ACTATGCAGCGTATAGCCCATAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Sphkap PubMed Links

Gene Details

Gene
Sphkap
Gene Name
SPHK1 interactor, AKAP domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172430.3 Intron NP_766018.3

View Full Product Details