Product Details

SNP ID
rs13459166
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:130541379 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATTCGGAACGCAAAGTGGATGTATA[C/T]GGGTTTGTGAGCCGGATCCGGGCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
4930473A02Rik PubMed Links

Gene Details

Gene
4930473A02Rik
Gene Name
RIKEN cDNA 4930473A02 gene
There are no transcripts associated with this gene.

Gene
Ptpra
Gene Name
protein tyrosine phosphatase, receptor type, A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163688.1 1673 Silent Mutation NP_001157160.1
NM_008980.2 1673 Silent Mutation NP_033006.2

View Full Product Details