Product Details
- SNP ID
-
rs13483048
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56697139 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCTTCAACCAGTGCAGGAGTG[A/G]GGCTGGATCTAGGCTGAGAGAGTCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ranbp3
PubMed Links
Gene Details
- Gene
- Ranbp3
- Gene Name
- RAN binding protein 3
View Full Product Details