Product Details
- SNP ID
-
rs13483552
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19097799 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAAAAGACTAGCACATCATAAGC[A/G]TTGATGTTACAGGTAGCGCTAATGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rorb
PubMed Links
Gene Details
- Gene
- Rorb
- Gene Name
- RAR-related orphan receptor beta
View Full Product Details