Product Details
- SNP ID
-
rs13482869
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:10584209 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTTGGGTGGGCTGTATCAACAGC[A/G]AGGAGTCAAGGTGAGCTGAGGCCTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pacrg
PubMed Links
Gene Details
- Gene
- Pacrg
- Gene Name
- PARK2 co-regulated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_027032.2 |
|
Intron |
|
|
NP_081308.1 |
View Full Product Details