Product Details

SNP ID
rs29312132
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:107612151 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTTTATTAACAATTTATTTCTTTA[A/C]TTAACTTCCCCTCCCTCCTCTCCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Ptprq PubMed Links

Gene Details

Gene
Ptprq
Gene Name
protein tyrosine phosphatase, receptor type, Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081432.1 Intron NP_001074901.1

View Full Product Details