Product Details

SNP ID
rs13476039
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:107607211 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGCCACTGCAGTGATTAGGAA[C/T]GCCCGGTGCTGTAGAACAGAACCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Serpinb8 PubMed Links

Gene Details

Gene
Serpinb8
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159748.1 1308 Intron NP_001153220.1
NM_011459.4 1308 Silent Mutation NP_035589.1

View Full Product Details